missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMD7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38137-20ul
This item is not returnable.
View return policy
Description
PSMD7 Polyclonal antibody specifically detects PSMD7 in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| PSMD7 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| 26S proteasome non-ATPase regulatory subunit 7, 26S proteasome regulatory subunit RPN8, Moloney leukemia virus-34 proviral integration, Mov34 homolog, Mov34 protein homolog, MOV3426S proteasome regulatory subunit S12, MOV34L, P40, proteasome (prosome, macropain) 26S subunit, non-ATPase, 7, proteasome (prosome, macropain) 26S subunit, non-ATPase, 7 (Mov34 homolog), Proteasome subunit p40, Rpn8, S12 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-324 of human PSMD7 (NP_002802.2).,, Sequence:, MPELAVQKVVVHPLVLLSVVDHFNRIGKVGNQKRVVGVLLGSWQKKVLDVSNSFAVPFDEDDKDDSVWFLDHDYLENMYGMFKKVNARERIVGWYHTGPKLHKNDIAINELMKRYCPNSVLVIIDVKPKDLGLPTEAYISVEEVHDDGTPTSKTFEHVTSEIGAEEAEEVGVEHLLRDIKDTTVGTLSQRITNQVHGLKGLNSKLLDIRSYLEKVATGKLPINHQIIYQLQDVFNLLPDVSLQEFVKAFYLKTNDQMVVVYLASLIRSVVALHNLINNKIANRDAEKKEGQEKEESKKDRKEDKEKDKDKEKSDVKKEEKKEKK | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 5713 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction