missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMD7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB120-11436
This item is not returnable.
View return policy
Description
PSMD7 Polyclonal specifically detects PSMD7 in Human, Mouse, Rat, Drosophila, Zebrafish samples. It is validated for Western Blot.
Specifications
| PSMD7 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 1mg/ml BSA with 0.05% Sodium Azide | |
| PSMD7 | |
| Synthetic peptide corresponding to residues A(55) V P F D E D D K D D S(66) C of human 19S subunit S12. | |
| 100 μg | |
| Primary | |
| Proteasome 19S S12 | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1 ug/ml | |
| 26S proteasome non-ATPase regulatory subunit 7, 26S proteasome regulatory subunit RPN8, Moloney leukemia virus-34 proviral integration, Mov34 homolog, Mov34 protein homolog, MOV3426S proteasome regulatory subunit S12, MOV34L, P40, proteasome (prosome, macropain) 26S subunit, non-ATPase, 7, proteasome (prosome, macropain) 26S subunit, non-ATPase, 7 (Mov34 homolog), Proteasome subunit p40, Rpn8, S12 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 5713 | |
| Human, Mouse, Rat, Drosophila, Zebrafish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction