missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PSMD8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552329
This item is not returnable.
View return policy
Description
Immunogen sequence: NRVAEFHTEL ERLPAKDIQT NVYIKHPVSL EQYLMEGSYN KVFLAKGNIP AESYTFFIDI LLDTIRDEIA GCIEKAYEKI LFTEATRILF FNTPKKMTDY AKKRGWVLGP NNYYSFASQQ QKPEDTTIPS TELAKQVIEY ARQ Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 97%.
The 26S proteasome is a multicatalytic proteinase complex with a highly ordered structure composed of 2 complexes, a 20S core and a 19S regulator. The 20S core is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. The 19S regulator is composed of a base, which contains 6 ATPase subunits and 2 non-ATPase subunits, and a lid, which contains up to 10 non-ATPase subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a non-ATPase subunit of the 19S regulator. A pseudogene has been identified on chromosome 1.
Specifications
| PSMD8 | |
| Polyclonal | |
| Unconjugated | |
| PSMD8 | |
| 26S proteasome non-ATPase regulatory subunit 8; 26S proteasome regulatory subunit p31; 26S proteasome regulatory subunit RPN12; 26S proteasome regulatory subunit S14; 6720456J22Rik; AA407360; AL033291; AL033322; AL033323; C76433; epididymis secretory sperm binding protein Li 91n; HEL-S-91n; HIP6; HYPF; Nin1p; p31; proteasome (prosome, macropain) 26S subunit, non-ATPase, 8; proteasome 26S non-ATPase subunit 8; proteasome 26S subunit, non-ATPase 8; proteasome 26S subunit, non-ATPase, 8; Psmd8; Rpn12; S14 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 292766, 5714, 57296 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P48556, Q9CX56 | |
| PSMD8 | |
| Recombinant protein corresponding to Human PSMD8. Recombinant protein control fragment (Product #RP-89240). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction