missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMF1 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98614C
This item is not returnable.
View return policy
Description
PSMF1 Polyclonal specifically detects PSMF1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| PSMF1 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 650 | |
| hPI31, PI31, proteasome (prosome, macropain) inhibitor subunit 1 (PI31), proteasome inhibitor hP131 subunit, proteasome inhibitor PI31 subunit | |
| Produced in rabbits immunized with E. coli-derived Human PSMF1 fragment. | |
| 0.1 mL | |
| Proteases & Other Enzymes | |
| 9491 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction