missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMF1 Antibody (OTI2D7), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73707DL594
This item is not returnable.
View return policy
Description
PSMF1 Monoclonal antibody specifically detects PSMF1 in Human, Rat samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunofluorescence
Specifications
| PSMF1 | |
| Monoclonal | |
| DyLight 594 | |
| hPI31, PI31, proteasome (prosome, macropain) inhibitor subunit 1 (PI31), proteasome inhibitor hP131 subunit, proteasome inhibitor PI31 subunit | |
| Full length recombinant protein of human PSMF1 (NP_848693) produced in HEK293T cells. | |
| 0.1 mL | |
| Proteases & Other Enzymes | |
| 9491 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Immunofluorescence | |
| OTI2D7 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction