missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMG2 Monoclonal Antibody (OTI2E12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808202S
This item is not returnable.
View return policy
Description
PSMG2 Monoclonal antibody specifically detects PSMG2 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| PSMG2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P21912 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 6390 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Immunohistochemistry (Paraffin) | |
| OTI2E12 | |
| Unconjugated | |
| Sdhb | |
| 1700017I17Rik; AW545363; CD40 ligand-activated specific transcript 3; CLAST3; HCCA3; HDCMC29P; hepatocellular carcinoma susceptibility protein; hepatocellular carcinoma-susceptibility protein 3; HSPC260; HsT1707; likely ortholog of mouse CD40 ligand-activated specific transcript 3 (Clast3); MDS003; PAC2; PAC-2; proteasome (prosome, macropain) assembly chaperone 2; proteasome a; proteasome assembling chaperone 2; proteasome assembly chaperone 2; PSMG2; TNFSF5IP1; Tumor necrosis factor superfamily member 5-induced protein 1; tumor necrosis factor superfamily, member 5-induced protein 1; x 003 protein | |
| Full length human recombinant protein of human SDHB produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction