missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMG2 Monoclonal Antibody (OTI5H6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808125
This item is not returnable.
View return policy
Description
Chaperone protein which promotes assembly of the 20S proteasome as part of a heterodimer with PSMG1. The PSMG1-PSMG2 heterodimer binds to the PSMA5 and PSMA7 proteasome subunits, promotes assembly of the proteasome alpha subunits into the heteroheptameric alpha ring and prevents alpha ring dimerization.Specifications
| PSMG2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q969U7 | |
| PSMG2 | |
| Full length human recombit protein of human PSMG2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI5H6 | |
| Unconjugated | |
| PSMG2 | |
| CLAST3, HCCA3, HsT1707, MDS003, PAC2, TNFSF5IP1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 56984 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction