missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMG4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-21369-100ul
This item is not returnable.
View return policy
Description
PSMG4 Polyclonal antibody specifically detects PSMG4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| PSMG4 | |
| Polyclonal | |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| proteasome assembly chaperone 4, bA506K6.2, C6orf86, PAC4, proteasome (prosome, macropain) assembly chaperone 4 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: DVSLHNFSARLWEQLVHFHVMRLTDSLFLWVGATPHLRNLAVAMCSRYDSIPVSTSLLGDTSDT | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 389362 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction