missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PSMG4 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5140040

Product Code. 17814560

  • € 464.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: LQNTDSNFAL LVENRIKEEM EAFPEKF Highest antigen sequence indentity to the following orthologs: Mouse - 89%, Rat - 89%.

Chaperone protein which promotes assembly of the 20S proteasome.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PSMG4
Polyclonal
Unconjugated
PSMG4
bA506K6.2; C6orf86; hPAC4; PAC4; PAC-4; proteasome (prosome, macropain) assembly chaperone 4; proteasome assembly chaperone 4; PSMG4
Rabbit
Antigen affinity chromatography
RUO
389362
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q5JS54
PSMG4
Recombinant protein fragment of Human PSMG4 (Product #RP-107593)
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.