missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSP/BPIFA2 Antibody (3F8E4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 187.00 - € 533.00
Specifications
| Antigen | PSP/BPIFA2 |
|---|---|
| Clone | 3F8E4 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18132099
|
Novus Biologicals
NBP2-37277 |
0.1 mL |
€ 533.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18660266
|
Novus Biologicals
NBP2-37277-0.025ml |
0.025 mL |
€ 187.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PSP/BPIFA2 Monoclonal specifically detects PSP/BPIFA2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
| PSP/BPIFA2 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
| Unconjugated | |
| RUO | |
| PBS with 0.05% Sodium Azide | |
| 140683 | |
| Purified recombinant fragment of mouse mSplunc2 (AA: 16-169) expressed in E. Coli. | |
| Primary | |
| 24.8 kDa |
| 3F8E4 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Human | |
| bA49G10.1, chromosome 20 open reading frame 70, Parotid secretory protein, PSP, short palate, lung and nasal epithelium carcinoma associated 2, short palate, lung and nasal epithelium carcinoma-associated protein 2, SPLUNC2 | |
| BPIFA2 | |
| IgG1 | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title