missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSP (REG1A) Monoclonal Antibody (OTI11E1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies CF813908
This item is not returnable.
View return policy
Description
PSP (REG1A) Monoclonal antibody specifically detects PSP (REG1A) in Human samples. It is validated for ELISASpecifications
| PSP (REG1A) | |
| Monoclonal | |
| See Label | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P05164 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 4353 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| ELISA | |
| OTI11E1 | |
| Unconjugated | |
| MPO | |
| ICRF; islet cells regeneration factor; Islet of Langerhans regenerating protein; LITHOST; Lithostathine; lithostathine 1 alpha; lithostathine-1-alpha; MGC12447; P19; pancreatic stone protein; pancreatic stone protein, secretory; pancreatic thread protein; protein-X; PSP; PSPS; PSPS1; PTP; RATLITHOST; RATRGPI; Reg; Reg1; REG1A; REG-1-alpha; regenerating family member 1 alpha; regenerating islet-derived 1 alpha; regenerating islet-derived mouse homolog 1; regenerating islet-derived protein 1-alpha; Regenerating protein I alpha; regeneration protein, lithostatin, pancreatic stone protein; Rgp1; RGPI | |
| Full length recombinant protein of human MPO (NP_000241) produced in Baculovirus-Insect cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction