missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSP94/MSMB Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99576-100ul
This item is not returnable.
View return policy
Description
PSP94/MSMB Polyclonal antibody specifically detects PSP94/MSMB in Mouse samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| PSP94/MSMB | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:500-1:2000 | |
| IGBF, MSMB, MSPB, PIP, PN44, PRPS, PSP57, PSP94 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human PSP94/MSMB | |
| 100 μg | |
| Cancer | |
| 4477 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction