missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSP94/MSMB Antibody (OTI3D5), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73625AF750
This item is not returnable.
View return policy
Description
PSP94/MSMB Monoclonal specifically detects PSP94/MSMB in Human samples. It is validated for Western Blot.
Specifications
| PSP94/MSMB | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| MSMB | |
| Purified prostate specific antigen (MSMB) from human seminal fluid. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2b |
| Western Blot | |
| OTI3D5 | |
| Western Blot | |
| IGBF, MSMB, MSPB, PIP, PN44, PRPS, PSP57, PSP94 | |
| Mouse | |
| Affinity Purified | |
| Cancer | |
| 4477.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction