Learn More
Invitrogen™ PSPH Recombinant Rabbit Monoclonal Antibody (1J5O2)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA555537
Description
Immunogen Sequence: MVSHSELRKL FYSADAVCFD VDSTVIREEG IDELAKICGV EDAVSEMTRR AMGGAVPFKA ALTERLALIQ PSREQVQRLI AEQPPHLTPG IRELVSRLQE RNVQVFLISG GFRSIVEHVA SKLNIPATNV FANRLKFYFN GEYAGFDETQ PTAESGGKGK VIKLLKEKFH FKKIIMIGDG ATDMEACPPA DAFIGFGGNV IRQQVKDNAK WYITDFVELL GELEE.
The protein encoded by this gene belongs to a subfamily of the phosphotransferases. This encoded enzyme is responsible for the third and last step in L-serine formation. It catalyzes magnesium-dependent hydrolysis of L-phosphoserine and is also involved in an exchange reaction between L-serine and L-phosphoserine. Deficiency of this protein is thought to be linked to Williams syndrome.
Specifications
| PSPH | |
| Recombinant Monoclonal | |
| 1.28 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| P78330, Q5M819, Q99LS3 | |
| PSPH | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-225 of human PSPH. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 1J5O2 | |
| Unconjugated | |
| PSPH | |
| AI480570; L-3-phosphoserine phosphatase; O-phosphoserine phosphohydrolase; OTTHUMP00000025059; Phosphoserine phosphatase; PSP; PSPase; Psph; PSPHD | |
| Rabbit | |
| Protein A | |
| RUO | |
| 100678, 304429, 5723 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.