missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSTPIP1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93547-0.02ml
This item is not returnable.
View return policy
Description
PSTPIP1 Polyclonal antibody specifically detects PSTPIP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| PSTPIP1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
| CD2BP1CD2-binding protein 1, CD2BP1L, CD2BP1S, H-PIPPEST phosphatase-interacting protein 1, PAPASCD2 antigen-binding protein 1, proline-serine-threonine phosphatase interacting protein 1, proline-serine-threonine phosphatase-interacting protein 1, PSTPIPCD2 cytoplasmic tail-binding protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 157-416 of human PSTPIP1 (NP_003969.2). AFERISANGHQKQVEKSQNKARQCKDSATEAERVYRQSIAQLEKVRAEWEQEHRTTCEAFQLQEFDRLTILRNALWVHSNQLSMQCVKDDELYEEVRLTLEGCSIDADIDSFIQAKSTGTEPPAPVPYQNYYDREVTPLTSSPGIQPSCGMIKRFSGLLHGSPKTTSLAASAASTETLTPTPERNEGVYTAIAVQEIQGNPASPAQEYRALYDYTAQNPDELDLSAGDILEVILEGEDGWWTVERNGQRGFVPGSYLEKL | |
| 0.02 mL | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 9051 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction