Learn More
IKA™ PT 100.51 Stainless Steel Temperature Sensor
Brand: IKA™ 0002600300
Additional Details : Weight : 0.16356kg
Specifications
8 mm | |
PT 100.51 | |
Temperature Sensor |
For RET Control 35193/94 | |
-20°C to +350°C |