missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTBP3 Mouse anti-Human, Clone: OTI5G7, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812031S
This item is not returnable.
View return policy
Description
PTBP3 Monoclonal antibody specifically detects PTBP3 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| PTBP3 | |
| Monoclonal | |
| 0.5-1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O95758 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 9991 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI5G7 | |
| Unconjugated | |
| PTBP3 | |
| 5830471K22Rik; AA407443; AI462022; AW107884; C86549; fission yeast differentiation regulator; polypyrimidine tract binding protein 3; polypyrimidine tract-binding protein 3; PTBP3; regulator of differentiation (in S. pombe) 1; regulator of differentiation 1; regulator of differentiation 1; polypyrimidine tract-binding protein 3; ROD1; ROD1 regulator of differentiation 1; si:dkey-27p1.2 | |
| Human recombinant protein fragment corresponding to amino acids 300-552 of human PTBP3 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction