missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PTCD2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110234
This item is not returnable.
View return policy
Description
Immunogen sequence: GKYKSALQVL IEMKNQDVKF TKDTYVLAFA ICYKLNSPES FKICTTLREE ALLKGEILSR RASCFAVALA LNQNEMAKAV SIFSQIMNPE SIACINLNII IHIQSNMLEN LIKTLKNAAE GNLSKFVKRH VFSEEVLAKV REKVKDVPAL VAKFDEIYGT LHITGQVTTD SLDAVLCHTP RDRKSHTLLL NKRMVSRRTF QPLSQSLLAE.
PTCD2 is a protein coding gene. Involved in mitochondrial RNA maturation and mitochondrial respiratory chain function.
Specifications
| PTCD2 | |
| Polyclonal | |
| Unconjugated | |
| PTCD2 | |
| 1190005P08Rik; CTC-365E16.1; FLJ12598; pentatricopeptide repeat domain 2; pentatricopeptide repeat-containing protein 2, mitochondrial; Ptcd2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 79810 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.64 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8WV60 | |
| PTCD2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 179-388 of human PTCD2 (NP_079030.3). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction