missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTCD3 Monoclonal Antibody (OTI1A8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies CF810262
This item is not returnable.
View return policy
Description
PTCD3 Monoclonal antibody specifically detects PTCD3 in Human samples. It is validated for Western BlotSpecifications
| PTCD3 | |
| Monoclonal | |
| See Label | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P40692 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 4292 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Western Blot | |
| OTI1A8 | |
| Unconjugated | |
| Mlh1 | |
| 2610034F17Rik; 2810422B04Rik; 28S ribosomal protein S39, mitochondrial; AA589622; AU045708; Mitochondrial small ribosomal subunit protein mS39; Mrps39; MRP-S39; pentatricopeptide repeat domain 3; pentatricopeptide repeat domain-containing protein 3, mitochondrial; pentatricopeptide repeat-containing protein 3, mitochondrial; Ptcd3; similar to CG4679-PA; transformation-related gene 15 protein; TRG15; TRG-15 | |
| Human recombinant protein fragment corresponding to amino acids 299-527 of human MLH1 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction