missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PTCRA Polyclonal Antibody
GREENER_CHOICE

Product Code. 15683415
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
400µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15683415 400 μL 400µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15683415 Supplier Invitrogen™ Supplier No. PA549122

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PTCRA Polyclonal Antibody for Western Blot

The pre-T-cell receptor complex (composed of PTCRA, TCRB and the CD3 complex) regulates early T-cell development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTCRA
Applications Western Blot
Classification Polyclonal
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene PTCRA
Gene Accession No. Q6ISU1
Gene Alias pre T-cell antigen receptor alpha; pre-T-cell receptor alpha chain; PTA; pT-alpha; pT-alpha-TCR; PTCRA
Gene Symbols PTCRA
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 197-225 amino acids from the C-terminal region of human PTCRA.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171558
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.