missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PTDSS2 Antibody (SR2476), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27752
This item is not returnable.
View return policy
Description
PTDSS2 Monoclonal antibody specifically detects PTDSS2 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| PTDSS2 | |
| Monoclonal | |
| Unconjugated | |
| 50 mM Tris-Glycine (pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 81490 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| SR2476 | |
| Western Blot 1:500-1:1000, Immunoprecipitation 1:20 | |
| phosphatidylserine synthase 2, serine-exchange enzyme II | |
| Recombinant protein of human PTDSS2 (Uniprot #: Q9BVG9) | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction