Learn More
PTEN Mouse anti-Human, CoraLite 594, Clone: 5C10B6, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL594-60300-100UL
Product Code. 16840916
  • € 450.00 / 100µL
Description
The PTEN antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PTEN. This antibody recognizes human, mouse antigen. The PTEN antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
Specifications
PTEN
Monoclonal
2000 μg/mL
Mouse
Primary
Human, Mouse
Immunofluorescence
5C10B6
CoraLite 594
100 μL
5728