Learn More
PTEN Mouse anti-Human, CoraLite 594, Clone: 5C10B6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL594-60300-100UL
Description
The PTEN antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PTEN. This antibody recognizes human, mouse antigen. The PTEN antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
| PTEN | |
| Monoclonal | |
| 2000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse |
| Immunofluorescence | |
| 5C10B6 | |
| CoraLite 594 | |
| 100 μL | |
| 5728 | |
| IgG2a |