missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PTEN2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA541332

Product Code. 15594772

  • € 530.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Peptide sequence: KILIDVFDGP PLYDDVKVQF FYSNLPTYYD NCSFYFWLHT SFIENNRLYL Sequence homology: Human: 100%.

TPTE encodes a putative transmembrane tyrosine phosphatase that may be involved in signal transduction pathways of the endocrine or spermatogenetic function of the testis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PTEN2
Polyclonal
Unconjugated
TPTE
Cancer/testis antigen 44; CT44; Pten2; PTEN-related tyrosine phosphatase; Putative tyrosine-protein phosphatase TPTE; tensin, putative protein-tyrosine phosphatase; TPTE; Transmembrane phosphatase with tensin homology; tumor antigen BJ-HCC-5
Rabbit
Protein A
RUO
7179
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Immunohistochemistry, Western Blot
1 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
P56180
TPTE
Synthetic peptide directed towards the C-terminal of human TPTE.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.