missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PTEN2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112778
This item is not returnable.
View return policy
Description
PTEN2 Polyclonal Antibody for Western Blot, ICC/IF, ELISA
TPTE encodes a putative transmembrane tyrosine phosphatase that may be involved in signal transduction pathways of the endocrine or spermatogenetic function of the testis.
Specifications
| PTEN2 | |
| Polyclonal | |
| Unconjugated | |
| TPTE | |
| Cancer/testis antigen 44; CT44; Pten2; PTEN-related tyrosine phosphatase; Putative tyrosine-protein phosphatase TPTE; tensin, putative protein-tyrosine phosphatase; TPTE; Transmembrane phosphatase with tensin homology; tumor antigen BJ-HCC-5 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 7179 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P56180 | |
| TPTE | |
| Recombinant Human Putative tyrosine-protein phosphatase TPTE protein (175-380AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction