Learn More
Poulten & Graf GmbH PTFE Adaptor for FORTUNA™ Dispenser with Thread
Product Code. 10249431
Click to view available options
Thread Size:
GL 25
GL 28
GL 30
GL 33
GL 36
GL 52
GL 55
GL 60
GL 65
Unit Size:
Each
Product Code. 10249431
Brand: Poulten & Graf GmbH 101 09125
Only null left
Add to basket
Add to basket
Complete a FORTUNA™ Dispenser with an adapter that comes in a wide selection of threads. Poulten & Graf GmbH PTFE Adaptor for FORTUNA™ Dispenser with Thread enables smaller containers to be supported on top the existing reservior.
  • Suitable for OPTIFIX and POLYFIX dispenser
  • Made of PTFE (Teflon™)
  • Adapter with threads
  • Can be used for acids and alkaline solutions that do not attack polypropylene
Specifications
Product Type Adapters for OPTIFIX and POLYFIX Dispensers
Autoclavable Autoclavable
Color White
Description Adapters for OPTIFIX and POLYFIX dispensers
Material PTFE
For Use With (Equipment) OPTIFIX and POLYFIX dispensers
Quantity 1
Thread Size GL 25