Learn More
Bohlender™ PTFE Cylindrical Power Magnetic Stirring Bar
Product Code. 10150092
missing translation for 'orderingAttributeHoverText'
Length (Metric):
50 mm
missing translation for 'unitSize'
Each
Product Code. 10150092
missing translation for 'mfr': Bohlender™ C36550
Only null left
Specifications
For Use With (Application) Used for agitating viscous liquids or for bridging larger distances between the magnetic stirring machine and the magnetic stirring bar
Diameter (Metric) 19 mm
Length (Metric) 50 mm
Material PTFE
Quantity 1/Pk.
Shape Cylinder