Learn More
Horst™ PTFE HST 42 Model Humidity-Proof Heating Cables
Specifications
Specifications
| Dimensions (L x W) | 13 Lm |
| Dimensions (L x W x H) | 13 Lm |
| Length (Metric) | 13 m |
| Electrical Requirements | 42V Voltage |
| Model | HST 42 |
| Length (Metric) Power Cord | 13m |
| Product Type | Humidity-proof Heating Cable |