Learn More
Horst™ PTFE HST Model Humidity-Proof Heating Cables
Click to view available options
Length (Metric) Power Cord:
1.1m
12m
5.5m
6m
8m
9m
Unit Size:
Each
Specifications
Specifications
| Dimensions (L x W) | 9 Lm |
| Dimensions (L x W x H) | 9 Lm |
| Electrical Requirements | ∼230V Voltage |
| Model | HST |
| Length (Metric) Power Cord | 9m |
| Product Type | Humidity-proof Heating Cable |