Learn More
Horst™ PTFE HST Model Humidity-Proof Heating Cables
Product Code. 10619812
Click to view available options
Length (Metric) Power Cord:
1.1m
12m
5.5m
6m
8m
9m
Unit Size:
Each
Product Code. 10619812
Brand: Horst™ 20821
Only null left
Specifications
Dimensions (L x W) 1.1 Lm
Dimensions (L x W x H) 1.1 Lm
Electrical Requirements ∼230V Voltage
Model HST
Length (Metric) Power Cord 1.1m
Product Type Humidity-proof Heating Cable