Learn More
Bel-Art™ SP Scienceware™ PTFE Magnetic Stirring Bar Kit
Shop All SP Scienceware Products
Click to view available options
Quantity:
3 sizes
3 sizes (2 each size)
7 sizes
Shape:
Octagon
Sphere
Unit Size:
Pack of 22
Pack of 6
Description
- Useful for environmental testing and life science applications in which small sample volumes need to be prepared and evaluated
- Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon™ PTFE low friction coating; durable and inert
- Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
- Autoclave safe
Specifications
Specifications
| Autoclavable | Autoclavable |
| Certifications/Compliance | ISO 9001:2008 |
| Dimensions (L x W x H) | 25.4 x 8, 38.1 x 8, 50.8 x 8mm |
| For Use With (Equipment) | Vials, Micro test tubes, Centrifuge tubes |
| Includes | Removable Pivot Rings |
| Coating | PTFE |
| Material | Alnico Magnet |
| Quantity | 3 sizes |
| Shape | Sphere |
| Type | Magnetic Stir Bar |