Learn More
Cole-Parmer™ PTFE Manifold, 4 outlets
Organize your fluid paths with these thermoplastic manifolds
Brand:  Cole-Parmer™ 31521-11
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11735268
  • € 181.00 / Each
Estimated Shipment
18-03-2021
Add to basket
Description
Description
  • PTFE Manifold.
  • Chemically-inert PTFE handles the most aggressive chemical solutions.
  • Available in a variety of configurations to suit a wide range of applications.
Specifications
Specifications
300°F
120 psi
4.75 in.
PTFE Manifold, 4 Outlets
-129°C
3/8 in. NPT(F)
3.81 cm
150°C
PTFE
12.07 cm
-200°F
1/2 in. NPT(F)
3.81 cm