Learn More
Cole-Parmer™ PTFE Manifold, 6 outlets
Organize your fluid paths with these thermoplastic manifolds
Brand:  Cole-Parmer™ 31521-15
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11755268
  • € 160.00 / Each
Estimated Shipment
27-04-2021
Add to basket
Description
Description
  • PTFE Manifold.
  • Chemically-inert PTFE handles the most aggressive chemical solutions.
  • Available in a variety of configurations to suit a wide range of applications.
Specifications
Specifications
300°F
120 psi
5 7/8 in.
PTFE Manifold, 6 Outlets
-129°C
1/4 in. NPT(F)
1.25 in.
1.25 in.
150°C
PTFE
14.93 cm
-200°F
3/8 in. NPT(F)
3.18 cm
3.18 cm