Learn More
VITLAB™ PTFE Polygonal Magnetic Stirring Bar
Click to view available options
Length (Metric):
5 mm
8 mm
10 mm
12 mm
15 mm
20 mm
25 mm
30 mm
40 mm
60 mm
70 mm
80 mm
Diameter (Metric):
2 mm
3 mm
4.5 mm
6 mm
7 mm
8 mm
10 mm
missing translation for 'unitSize'
Pack of 10
Description
- Permanent magnet AlNiCo V core
- Angled shape gives rise to significant turbulence
- Achieves effective mixing, even at low spin rates
- FTE
Specifications
Specifications
| For Use With (Application) | For preparation, separation and concentration |
| Includes | Permanent magnet AlNiCo V core |
| Diameter (Metric) | 6 mm |
| Length (Metric) | 30 mm |
| Material | PTFE |
| Quantity | 10/Pk. |
| Shape | Polygon |