Learn More
Amarell™ AD20TH Digital Thermometer Probe
Description
- Precise Pt100 class A probe
- Stainless steel
- PVC cable, 1m long
Specifications
Specifications
| Diameter (Metric) Probe | 3 mm |
| For Use With (Application) | For gases, liquids, soft plastic materials |
| Length (Metric) Cable | 1 m |
| Length (Metric) Probe | 105 mm |
| Material | Stainless Steel, PVC |
| Measurement Time | T90 = 5 sec. |
| Probe Type | Immersion Probe |
| Max. Temperature (Metric) | 300°C |
| Min. Temperature (Metric) | -50°C |
| Temperature Range (Metric) | -50°C to +300°C |
| Show More |