Learn More
Duran™ PTFE Valve Pins
Product Code. 10235969 Shop All DWK Life Sciences Products
Click to view available options
Description:
Passage Size: 0 to 4mm
Passage Size: 0 to 6mm
Unit Size:
Each
Product Code. 10235969
Brand: Duran™ 286057207
Specifications
Type Valve Pin
Description Passage Size: 0 to 6mm
Material PTFE
For Use With (Application) For Produran-valve tap
Includes With Abdrehsicherung