Learn More
Duran™ PTFE Valve Pins
Shop All DWK Life Sciences Products
missing translation for 'orderingAttributeHoverText'
Description:
Passage Size: 0 to 4mm
Passage Size: 0 to 6mm
missing translation for 'unitSize'
Each
Specifications
Specifications
| Type | Valve Pin |
| Description | Passage Size: 0 to 4mm |
| Material | PTFE |
| For Use With (Application) | For Produran-valve tap |
| Includes | With Abdrehsicherung |