Learn More
Kartell™ PTFE Watch Glass
Product Code. 10224141
Click to view available options
Diameter (Metric):
52.76 mm
76.32 mm
99.39 mm
126.36 mm
153.92 mm
Unit Size:
Pack of 2
Product Code. 10224141
Brand: Kartell™ 0064700
Only null left
Add to basket
Add to basket
Use a watch glass to evaporate liquids, hold solids when weighing a sample, and in situations where cross-contamination could occur. Kartell™ PTFE Watch Glass has good heat and chemical resistance.
  • Superior chemical, mechanical, and heat resistance
  • Can be used safely for continuous service at temperatures up to 250°C
  • Suitable for foodstuff
Specifications
Bottom Shape Round
Material PTFE
Shape Round
Type Watch Glass
Diameter (Metric) 153.92 mm