missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTGDR Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NLS8133
This item is not returnable.
View return policy
Description
PTGDR Polyclonal antibody specifically detects PTGDR in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| PTGDR | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 3 - 13 μg/mL | |
| AS1, ASRT1, DP, DP1, MGC49004, PGD receptor, PGD2 receptor, prostaglandin D2 receptor, prostaglandin D2 receptor (DP), Prostanoid DP receptor | |
| Synthetic 20 amino acid peptide from 3rd cytoplasmic domain of human PTGDR. | |
| 0.05 mL | |
| GPCR | |
| 5729 | |
| Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction