missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PTGDR Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102298
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total PTGDR.
The protein encoded by this gene is a G-protein-coupled receptor. It has been shown to function as a prostanoid DP receptor. The activity of this receptor is mainly mediated by G-S proteins that stimulate adenylate cyclase resulting in an elevation of intracellular cAMP and Ca2+. Knockout studies in mice suggest that the ligand of this receptor, prostaglandin D2 (PGD2), functions as a mast cell-derived mediator to trigger asthmatic responses.
Specifications
| PTGDR | |
| Polyclonal | |
| Unconjugated | |
| PTGDR | |
| AS1; ASRT1; DP; Dp receptor; DP1; MGC49004; PGD receptor; PGD2 receptor; prostaglandin D2 receptor; prostaglandin D2 receptor (DP); Prostanoid DP receptor; PTGDR; PTGDR1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 5729 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q13258 | |
| PTGDR | |
| A synthesized peptide derived from human PTGDR(Accession Q13258), corresponding to amino acid residues A268-V318. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction