missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTGER2 Antibody (SR1927), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21756-100ul
This item is not returnable.
View return policy
Description
PTGER2 Monoclonal antibody specifically detects PTGER2 in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence
Specifications
| PTGER2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Bovine | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence | |
| SR1927 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| EP2, PGE receptor EP2 subtype, PGE2 receptor EP2 subtype, prostaglandin E receptor 2 (subtype EP2), 53kD, prostaglandin E receptor 2 (subtype EP2), 53kDa, prostaglandin E2 receptor EP2 subtype, Prostanoid EP2 receptor | |
| A synthesized peptide derived from human PTGER2 (Uniprot #: P43116) | |
| 100 μg | |
| Cancer, GPCR, Signal Transduction | |
| 5732 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction