missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTGES3 Monoclonal Antibody (OTI2F2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803290S
This item is not returnable.
View return policy
Description
PTGES3 Monoclonal antibody specifically detects PTGES3 in Human samples. It is validated for Western BlotSpecifications
| PTGES3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q15185 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10728 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI2F2 | |
| Unconjugated | |
| Ptges3 | |
| 5730442A20Rik; Co chaperone p23; cPGES; cytosolic prostaglandin E synthase; cytosolic prostaglandin E2 synthase; Hsp90 co chaperone; Hsp90 co-chaperone; LOC100351480; P23; p23 cochaperone; progesterone receptor; progesterone receptor complex p23; Prostaglandin E synthase 3; prostaglandin E synthase 3 (cytosolic); prostaglandin-E synthase 3; Ptges; PTGES3; ptges3 {ECO:0000250; RGD1561913; sid 3177; Sid3177; Tebp; telomerase binding protein, p23; telomerase-binding protein p23; unactive progesterone receptor, 23 kD; UniProtKB:Q15185} | |
| Full length human recombinant protein of human PTGES3 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction