Learn More
PTGES3 Monoclonal Antibody (OTI2H10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA803436
Encompass_Preferred
 View more versions of this product
Product Code. 17542503
  • POA / 100µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
Cytosolic prostaglandin synthase that catalyzes the oxidoreduction of prostaglandin endoperoxide H2 (PGH2) to prostaglandin E2 (PGE2). Molecular chaperone that localizes to genomic response elements in a hormone-dependent manner and disrupts receptor-mediated transcriptional activation, by promoting disassembly of transcriptional regulatory complexes. Facilitates HIF alpha proteins hydroxylation via interaction with EGLN1/PHD2, leading to recruit EGLN1/PHD2 to the HSP90 pathway.
Specifications
Specifications
PTGES3
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q15185
PTGES3
Full length human recombit protein of human PTGES3 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG2a
Western Blot
OTI2H10
Unconjugated
PTGES3
P23, TEBP, cPGES
Mouse
Affinity Chromatography
RUO
10728
-20° C, Avoid Freeze/Thaw Cycles