missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTGFR Antibody (1014602) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB10294JF525
This item is not returnable.
View return policy
Description
PTGFR Monoclonal antibody specifically detects PTGFR in Human samples. It is validated for Flow Cytometry,CyTOF
Specifications
| PTGFR | |
| Monoclonal | |
| Janelia Fluor 525 | |
| FP, MGC120498, MGC46203, PGF receptor, PGF2 alpha receptor, PGF2-alpha receptor, prostaglandin F receptor (FP), prostaglandin F2 alpha receptor, prostaglandin F2-alpha receptor, prostaglandin receptor (2-alpha), Prostanoid FP receptor | |
| Synthetic peptide containing human PTGFR RD3 epitope, Accession # P43088 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, CyTOF | |
| 1014602 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| GPCR | |
| 5737 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction