missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PTGFR Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534128
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Horse (100%) Mouse, Rat, Dog (88%).
The protein encoded by this gene is member of the G-protein coupled receptor family. This protein is a receptor for prostaglandin F2-alpha (PGF2-alpha), which is known to be a potent luteolytic agent, and may also be involved in modulating intraocular pressure and smooth muscle contraction in uterus. Knockout studies in mice suggest that the interaction of PGF2-alpha with this receptor may initiate parturition in ovarian luteal cells and thus induce luteolysis. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
| PTGFR | |
| Polyclonal | |
| Unconjugated | |
| PTGFR | |
| AI957154; fp; FP prostanoid receptor; FP prostanoid receptor FPB isoform; FP receptor; PF2AR; PGF; PGF receptor; PGF2 alpha receptor; PGF2-alpha receptor; prostaglandin F receptor; prostaglandin F receptor (FP); prostaglandin F2 alpha receptor; prostaglandin F2alpha receptor; prostaglandin F2-alpha receptor; prostaglandin F2alpha receptor FPa isoform; prostaglandin F2alpha receptor isoform-aplpha; prostaglandin F2alpha receptor isoform-beta; prostaglandin F2alpha receptor isoform-delta; prostaglandin F2alpha receptor isoform-epsiron; prostaglandin F2alpha receptor isoform-gamma; prostaglandin receptor (2-alpha); prostanoid FP receptor; Ptgfr | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100009714, 5737 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| P43088 | |
| PTGFR | |
| Synthetic 16 amino acid peptide from 2nd extracellular domain of human PTGFR. | |
| 50 μg | |
| Primary | |
| Human, Monkey, Horse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction