Learn More
Invitrogen™ PTGIR Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5102240
Product Code. 16648714
  • € 491.00 / 100µL
Description
Antibody detects endogenous levels of total Prostacyclin Receptor.
Receptor for prostacyclin (prostaglandin I2 or PGI2). The activity of this receptor is mediated by G(s) proteins which activate adenylate cyclase.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PTGIR
Polyclonal
Unconjugated
PTGIR
IP; IP receptor; PGI receptor; PGI2; PGI2 receptor; PRIPR; Prostacyclin receptor; prostaglandin I receptor (IP); prostaglandin I2 (prostacyclin) receptor (IP); prostaglandin I2 receptor; Prostanoid IP receptor; Ptgir
Rabbit
Affinity Chromatography
RUO
5739
-20°C
Liquid
Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P43119
PTGIR
A synthesized peptide derived from human PTGIR(Accession P43119), corresponding to amino acid residues G204-P254.