Learn More
Invitrogen™ PTGIR Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102240
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total Prostacyclin Receptor.
Receptor for prostacyclin (prostaglandin I2 or PGI2). The activity of this receptor is mediated by G(s) proteins which activate adenylate cyclase.
Specifications
| PTGIR | |
| Polyclonal | |
| Unconjugated | |
| PTGIR | |
| IP; IP receptor; PGI receptor; PGI2; PGI2 receptor; PRIPR; Prostacyclin receptor; prostaglandin I receptor (IP); prostaglandin I2 (prostacyclin) receptor (IP); prostaglandin I2 receptor; Prostanoid IP receptor; Ptgir | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 5739 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P43119 | |
| PTGIR | |
| A synthesized peptide derived from human PTGIR(Accession P43119), corresponding to amino acid residues G204-P254. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |