Learn More
PTGIS Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27061-1-AP-20UL
Description
The PTGIS antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PTGIS. This antibody recognizes human, mouse antigen. The PTGIS antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| PTGIS | |
| 550 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG |
| Western Blot, Immunofluorescence, Immunohistochemistry, ELISA | |
| CYP8, CYP8A1, PGIS, Prostacyclin synthase, Prostaglandin I2 synthase, PTGI, PTGIS | |
| 20 μL | |
| 5740 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |