missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PTGR1 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5143286
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
Additional Information: This antibody is expected to recognize both reported isoforms (NP_036344.2; NP_001139581.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.
PTGR1 encodes an enzyme that is involved in the inactivation of the chemotactic factor, leukotriene B4. The encoded protein specifically catalyzes the NADP+ dependent conversion of leukotriene B4 to 12-oxo-leukotriene B4. A pseudogene of this gene is found on chromosome 1. Alternative splicing results in multiple transcript variants.
Tekniske data
| PTGR1 | |
| Polyclonal | |
| Unconjugated | |
| PTGR1 | |
| 12-HD; 15-oxoprostaglandin 13-reductase; 2510002C21Rik; ADRAB-F; D3T-inducible gene 1 protein; Dig1; DIG-1; Dithiolethione-inducible gene 1 protein; dithiolethione-inducible gene-1; FLJ99229; leukotriene B4 12-hydroxydehydrogenase; leukotriene b4 12-hydroxydehydrogenase/prostaglandin 15-keto reductase; LTB4; LTB4DH; MGC34943; NADP-dependent leukotriene B4 12-hydroxydehydrogenase; PGR; PGR1; PRG-1; Prostaglandin reductase 1; PTGR1; unnamed protein product; Zadh3; zinc binding alcohol dehydrogenase domain containing 3 | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 22949 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q14914 | |
| PTGR1 | |
| Peptide with sequence C-PTNSDFELKTAELP. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion