missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PTGR2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15799470 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15799470 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15799470 Supplier Invitrogen™ Supplier No. PA551518

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: VAGKIGHFLG CSRVVGICGT HEKCILLTSE LGFDAAINYK KDNVAEQLRE SCPAGVDVYF DNVGGNISDT VISQMNENSH IILCGQISQY NKDVPYPPPL SPAIEAIQKE RNITRERFLV LNYKDKFEPG ILQLSQWFKE Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 87%.

ZADH1 (zinc binding alcohol dehydrogenase, domain containing 1), also known as PTGR2 (prostaglandin reductase 2), PGR2 or 15-oxoprostaglandin 13-reductase, is a 351 amino acid cytoplasmic protein that belongs to the NADP-dependent oxidoreductase L4BD family. Functioning as s a 15-oxoprostaglandin 13-reductase, ZADH1 catalyzes the conversion of 15-keto-prostaglandin E2 to 15-keto-13, 14-dihydro-prostaglandin E2 in a NADPH-dependent manner. ZADH1 overexpression has been found to repress PPAR gamma transcriptional activity and adipocyte differentiation. Widely expressed, ZADH1 is found at highest levels in heart, kidney, liver, pancreas and prostate, with moderate levels found in brain, small intestine, lung, testis and skeletal muscle. ZADH1 exists as a monomer, utilizes NADPH as a cofactor, and undergoes alternative splicing to produce two isoforms that are encoded by a gene located on human chromosome 14q24.3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTGR2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PTGR2
Gene Accession No. Q8N8N7
Gene Alias 15-oxoprostaglandin 13-reductase; 15-oxoprostaglandin 13-reductase {ECO:0000250; 15-oxoprostaglandin-delta13-reductase; 1810016I24Rik; 9130222H03Rik; 9630002F03Rik; AI838763; B830026H24Rik; epididymis secretory protein Li 298; HEL-S-298; PGR2; PGR-2; PRG2; PRG-2; PRG-2 {ECO:0000250; prostaglandin reductase 2; prostaglandin reductase 2 {ECO:0000250; PTGR2; ptgr2 {ECO:0000250; UniProtKB:Q8VDQ1}; ZADH1; zinc binding alcohol dehydrogenase, domain containing 1; zinc-binding alcohol dehydrogenase domain-containing protein 1
Gene Symbols PTGR2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PTGR2. Recombinant protein control fragment (Product #RP-88892).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 145482
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.