missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTGS2 Monoclonal Antibody (OTI2E1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA805370S
This item is not returnable.
View return policy
Description
PTGS2 Monoclonal antibody specifically detects PTGS2 in Human samples. It is validated for Western BlotSpecifications
| PTGS2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q14994 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 9970 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI2E1 | |
| Unconjugated | |
| NR1I3 | |
| COII; COX2; Cox-2; COXII; cyclooxygenase; cyclooxygenase 2; cyclooxygenase 2b; cyclooxygenase-2; Cytochrome c oxidase polypeptide II; Cytochrome c oxidase subunit 2; cytochrome c oxidase subunit II; glucocorticoid-regulated inflammatory cyclooxygenase; Gripghs; hCox-2; Macrophage activation-associated marker protein P71/73; mitochondrially encoded cytochrome c oxidase II; MTCO2; MT-CO2; PES-2; PGG/HS; PGH synthase 2; Pghs2; PGHS-2; Pghs-b; PHS II; PHS-2; prostaglandin G/H synthase 2; Prostaglandin H2 synthase 2; prostaglandin-endoperoxide synthase 2; prostaglandin-endoperoxide synthase 2 (prostaglandin G/H synthase and cyclooxygenase); Ptgs2; Tis10; TIS10 protein | |
| Human recombinant protein fragment corresponding to amino acids 1-236 of human NR1I3 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction