missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTGS2 Monoclonal Antibody (OTI9H10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA805365
This item is not returnable.
View return policy
Description
Prostaglandin-endoperoxide synthase (PTGS), also known as cyclooxygenase, is the key enzyme in prostaglandin biosynthesis, and acts both as a dioxygenase and as a peroxidase. There are two isozymes of PTGS: a constitutive PTGS1 and an inducible PTGS2, which differ in their regulation of expression and tissue distribution. This gene encodes the inducible isozyme. It is regulated by specific stimulatory events, suggesting that it is responsible for the prostanoid biosynthesis involved in inflammation and mitogenesis.Specifications
| PTGS2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P35354 | |
| PTGS2 | |
| Human recombit protein fragment corresponding to amino acids 18-260 of human PTGS2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI9H10 | |
| Unconjugated | |
| PTGS2 | |
| COX-2, COX2, GRIPGHS, PGG/HS, PGHS-2, PHS-2, hCox-2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5743 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction